5-Methylpyridin-3-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

448740010 448740050 448742500
TS448740010EA 186 USD
TS448740010 TS448740050 TS448742500
5-Methylpyridin-3-ol 97%
5-Methylpyridin-3-ol
Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now

Specification Test Results

Appearance Light yellow to brown Powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
GC ≥96.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR