Lead(II) perchlorate trihydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead diperchlorate trihydrate

447781000 447785000
TS447781000EA 83 USD
TS447781000 TS447785000
Lead(II) perchlorate trihydrate ACS
Lead(II) perchlorate trihydrate
Formula: Cl₂O₈Pb·3H₂O
MW: 460.15 g/mol
Melting Pt: 280 (dec.)
Density: 2.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149817
CAS Number: 13453-62-8
UN: 1470
ADR: 5.1 II

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric 97.0 to 102.0 %
pH 3.0 to 5.0 (5% in water, 25.0°C)
Chloride (Cl) ≤0.01 %
Iron (Fe) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR