2-Morpholino-4-pyridineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine

445200010 445200050
TS445200010EA 258 USD
TS445200010 TS445200050
2-Morpholino-4-pyridineboronic acid pinacol ester 97%
2-Morpholino-4-pyridineboronic acid pinacol ester
Formula: C₁₅H₂₃BN₂O₃
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD11054039
CAS Number: 888721-86-6

Order Now

Specification Test Results

Appearance White to off-white to light brown Powder or solid
Infrared spectrum Conforms
Melting point 82 to 88 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR