2-Amino-5-chloropyrimidine 97%

Supplier: Thermo Scientific Chemicals
Danger

443760010 443760050
TS443760010EA 20 USD
TS443760010 TS443760050
2-Amino-5-chloropyrimidine 97%
2-Amino-5-chloropyrimidine
Formula: C₄H₄ClN₃
Boiling Pt: 314 °C (760 mmHg)
Melting Pt: 235…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014604
CAS Number: 5428-89-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR