5-Bromo-2-methoxypyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

441850010 441850050
TS441850010EA 108 USD
TS441850010 TS441850050
5-Bromo-2-methoxypyrimidine 97%
5-Bromo-2-methoxypyrimidine
Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD04973351
CAS Number: 14001-66-2

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR