4-Cyanopiperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Piperidine-4-carbonitrile

441820050 441820250
TS441820050EA 72 USD
TS441820050 TS441820250
4-Cyanopiperidine 97%
4-Cyanopiperidine
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 100…105 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD05022468
CAS Number: 4395-98-6
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR