4-Cyanophenylacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

437730010 437730050
TS437730010EA 122 USD
TS437730010 TS437730050
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid
Formula: C₉H₇NO₂
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06798066
CAS Number: 5462-71-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150.0 to 155.0 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR