Silicon dioxide 99.998% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

437150050 437150200 437151000
TS437150050EA 48 USD
TS437150050 TS437150200 TS437151000
Silicon dioxide 99.998% (trace metals basis)
Silicon dioxide
Formula: SiO₂
Melting Pt: 1610 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Assay 44.5 to 47.9 % (Si)
Total trace metal impurities ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR