Tellurium 99.999% (trace metals basis), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
437010250
437011000
TS437010250EA
115
USD
TS437010250
TS437011000
Tellurium 99.999% (trace metals basis), powder
Tellurium
Formula:
Te MW: 127.6 g/mol Boiling Pt: 990 °C (760 mmHg) Melting Pt: 450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 |
Specification Test Results
Appearance | Grey to black Powder |
Total trace metal impurities | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...