9-Fluorenylmethyl carbamate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbamic Acid 9-Fluorenylmethyl Ester, Fmoc-NH2, (9H-Fluoren-9-ylmethoxy)carboxamide

434170010 434170050
TS434170010EA 104 USD
TS434170010 TS434170050
9-Fluorenylmethyl carbamate 99%
9-Fluorenylmethyl carbamate
Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Melting Pt: 204…207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00237376
CAS Number: 84418-43-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR