Methyl-6-chloropyridazine-4-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

432980010 432980050
TS432980010EA 234 USD
TS432980010 TS432980050
Methyl-6-chloropyridazine-4-carboxylate 97%
Methyl-6-chloropyridazine-4-carboxylate
Formula: C₆H₅ClN₂O₂
Melting Pt: 88.4…88.5 °C
Storage Temperature: Ambient
MDL Number: MFCD16660968
CAS Number: 1093860-48-0

Order Now

Specification Test Results

Appearance Cream to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 81 to 89 °C
GC ≥96.0 %
UV at 230 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.18 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR