Azetidin-2-one 98%

Supplier: Thermo Scientific Chemicals
Danger

432960050 432960010
TS432960050EA 460 USD
TS432960050 TS432960010
Azetidin-2-one 98%
Azetidin-2-one
Formula: C₃H₅NO
Boiling Pt: 106 °C (15 mmHg)
Melting Pt: 72.5…76.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013328
CAS Number: 930-21-2
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 72 to 77 °C
GC ≥97.5 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR