4-Nitrophenyl trifluoromethanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

425790010 425790050
TS425790010EA 24 USD
TS425790010 TS425790050
4-Nitrophenyl trifluoromethanesulfonate 99%
4-Nitrophenyl trifluoromethanesulfonate
Formula: C₇H₄F₃NO₅S
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00192354
CAS Number: 17763-80-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 52 to 55 °C
GC ≥98.5 %
Iron (Fe) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR