Ethyl picolinoylacetate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-oxo-3-(2-pyridyl)propionate, Ethyl 3-oxo-3-(pyridin-2-yl)propanoate

425470025 425470100
TS425470025EA 140 USD
TS425470025 TS425470100
Ethyl picolinoylacetate 95%
Ethyl picolinoylacetate
Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Boiling Pt: 100…104 °C (.3 mmHg)
Density: 1.164 g/cm³
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00094022
CAS Number: 26510-52-1

Order Now

Specification Test Results

Appearance Clear straw yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5190 (20°C, 589 nm)
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR