Sodium diphenylamine-4-sulfonate ACS

Supplier: Thermo Scientific Chemicals
Warning

423660500 423660010 423660050
TS423660500EA 313 USD
TS423660500 TS423660010 TS423660050
Sodium diphenylamine-4-sulfonate ACS
Sodium diphenylamine-4-sulphonate
Formula: C₁₂H₁₀NNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Order Now

Specification Test Results

Appearance Off-white to beige to light grey Crystalline powder
Infrared spectrum Conforms
Sensitivity (as indicator) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR