Tin ACS 30 mesh
Supplier: Thermo Scientific Chemicals
420995000
420991000
TS420995000EA
267
USD
TS420995000
TS420991000
Tin ACS 30 mesh
Tin
Formula:
Sn Boiling Pt: 2270 °C (760 mmHg) Melting Pt: 231.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 |
Specification Test Results
Appearance | Grey Metallic granular powder |
Titration Complexometric | ≥99.5 % |
Antimony (Sb) | ≤200 ppm |
Arsenic (As) | ≤1 ppm |
Copper (Cu) | ≤50 ppm |
Iron (Fe) | ≤100 ppm |
Lead (Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...