Tellurium 99+% (trace metals basis), powder

Supplier: Thermo Scientific Chemicals
Warning

420020100 420021000
TS420020100EA 31 USD
TS420020100 TS420021000
Tellurium 99+% (trace metals basis), powder
Tellurium
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990
Melting Pt: 450
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
Merck Index: 12,09273

Order Now

Specification Test Results

Appearance Grey to black Powder
Total trace metal impurities ≤1.0 %
Melting point 77 to 81 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR