Disodium metasilicate nonahydrate 44-47.5% (total solids)

Supplier: Thermo Scientific Chemicals
Danger

419590025 419590250 419595000
TS419590025EA 396 USD
TS419590025 TS419590250 TS419595000
Disodium metasilicate nonahydrate 44-47.5% (total solids)
Disodium metasilicate nonahydrate
Formula: Na₂SiO₃·9H₂O
Melting Pt: 72 °C
Storage Temperature: Ambient
CAS Number: 13517-24-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III

Order Now

Specification Test Results

Appearance White Granular powder or granules (May have a yellow-pink hue)
Assay 44 to 47.5 % total solids
Ratio 1.02 to 1.06 molar ratio SiO2/Na2O
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR