Polyethylene glycol 8,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

418050010 418050030 418050100
TS418050010EA 116 USD
TS418050010 TS418050030 TS418050100
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250
Melting Pt: 55…60 °C
Flash Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Hydroxyl value 12 to 16 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR