4-Amino-2-chloro-5-fluoropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

399980010 399980050
TS399980010EA 243 USD
TS399980010 TS399980050
4-Amino-2-chloro-5-fluoropyrimidine 98%
4-Amino-2-chloro-5-fluoropyrimidine
Formula: C₄H₃ClFN₃
MW: 147.54 g/mol
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00057344
CAS Number: 155-10-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR