4-Hydroxyindanone 97%

Supplier: Thermo Scientific Chemicals
Warning

399030010 399030050
TS399030010EA 108 USD
TS399030010 TS399030050
4-Hydroxyindanone 97%
4-Hydroxyindanone
Formula: C₉H₈O₂
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00143330
CAS Number: 40731-98-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 241 to 244 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR