2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde, 1,4-Benzodioxan-6-carboxaldehyde, 2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
398210010
398210050
TS398210010EA
36
USD
TS398210010
TS398210050
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 98%
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
Formula:
C₉H₈O₃ MW: 164.16 g/mol Boiling Pt: 105 °C (20hPa) Melting Pt: 52…53 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010092 CAS Number: 29668-44-8 |
Specification Test Results
Appearance | Yellow to light beige Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 48 to 52 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...