3',5'-Difluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one

398120010 398120050
TS398120010EA 60 USD
TS398120010 TS398120050
3',5'-Difluoroacetophenone 98%
3',5'-Difluoroacetophenone
Formula: C₈H₆F₂O
MW: 156.13 g/mol
Melting Pt: 34…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00042489
CAS Number: 123577-99-1

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR