4-Amino-2-methylpyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine

393650010 393650050
TS393650010EA 87 USD
TS393650010 TS393650050
4-Amino-2-methylpyridine 98+%
4-Amino-2-methylpyridine
Formula: C₆H₈N₂
MW: 108.14
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00186509
CAS Number: 18437-58-6

Order Now

Specification Test Results

Appearance Tan Powder
Infrared spectrum Conforms
Melting point 92 to 95 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR