Methyl trans-3-hexenoate predominantly trans 95+%

Supplier: Thermo Scientific Chemicals
Warning

393550050 393550250
TS393550050EA 19 USD
TS393550050 TS393550250
Methyl trans-3-hexenoate predominantly trans 95+%
Methyl trans-3-hexenoate
Formula: C₇H₁₂O₂
Boiling Pt: 169 °C (760 mmHg)
Density: 0.915 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
CAS Number: 2396-78-3
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20 °C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR