(R)-1-(pyridin-4-yl)ethanol 99+%, (99+% ee)

Supplier: Thermo Scientific Chemicals
Warning

393362500 393360010
TS393362500EA 87 USD
TS393362500 TS393360010
(R)-1-(pyridin-4-yl)ethanol 99+%, (99+% ee)
(R)-1-(pyridin-4-yl)ethanol
Formula: C₇H₉NO
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077865
CAS Number: 27854-88-2

Order Now

Specification Test Results

Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥99.0 %
Enantiomeric excess ≥99.0 %
Specific optical rotation +53° to +59° (20 °C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR