3,5-Dibromoanthranilic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
393160010
393160050
393160250
TS393160010EA
11
USD
TS393160010
TS393160050
TS393160250
3,5-Dibromoanthranilic acid 98+%
3,5-Dibromoanthranilic acid
Formula:
C₇H₅Br₂NO₂ Melting Pt: 235…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017087 CAS Number: 609-85-8 |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...