3-(4-Pyridyl)acrylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

391750010 391750050
TS391750010EA 59 USD
TS391750010 TS391750050
3-(4-Pyridyl)acrylic acid 97%
3-(4-Pyridyl)acrylic acid
Formula: C₈H₇NO₂
Melting Pt: 280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040746
CAS Number: 5337-79-1

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR