3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

391000010 391000050
TS391000010EA 30 USD
TS391000010 TS391000050
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97+%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Formula: C₁₁H₉NO₄
Melting Pt: 149…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00023096
CAS Number: 3339-73-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Titration with NaOH ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR