Sodium 99.8%, oiled sticks, wrapped in aluminium foil

Supplier: Thermo Scientific Chemicals
Danger

390120010 390121000 390122500
TS390120010EA 186 USD
TS390120010 TS390121000 TS390122500
Sodium 99.8%, oiled sticks, wrapped in aluminium foil
Sodium
Formula: Na
Boiling Pt: 883 °C (760 mmHg)
Melting Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00085307
CAS Number: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 12,08710

Order Now

Specification Test Results

Appearance Grey Sticks
Calcium (Ca) ≤400 ppm
Potassium (K) ≤300 ppm
Sodium (Na) ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR