4-Bromobenzaldehyde diethyl acetal 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-α,α-diethoxytoluene, 1-Bromo-4-(diethoxymethyl)benzene

390020100 390020500
TS390020100EA 31 USD
TS390020100 TS390020500
4-Bromobenzaldehyde diethyl acetal 99%
4-Bromobenzaldehyde diethyl acetal
Formula: C₁₁H₁₅BrO₂
Boiling Pt: 75 °C (.2 mmHg)
Density: 1.274 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD01863514
CAS Number: 34421-94-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5120 to 1.5140 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR