Calcium acetylide 72-82%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Calcium acetylide

389792500 389790010 389790025
TS389792500EA 33 USD
TS389792500 TS389790010 TS389790025
Calcium acetylide 72-82%
Calcium acetylide
Formula: CaC₂
Boiling Pt: 2300 °C (760 mmHg)
Melting Pt: 2160 °C
Storage Temperature: Ambient
MDL Number: MFCD00010905
CAS Number: 75-20-7
UN: 1402
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Dark grey Lumps
Titration Complexometric 12 to 23 % (CaO)
Particle size 7 to 25 mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR