Wood's alloy (lipowitz's alloy), rods
Supplier: Thermo Scientific Chemicals
|
Danger
|
388551000
388555000
TS388551000EA
58
USD
TS388551000
TS388555000
Wood's alloy (lipowitz's alloy), rods
Wood's alloy (lipowitz's alloy)
Melting Pt:
70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00198175 CAS Number: 76093-98-6 UN: 2570 ADR: 6.1,II |
Specification Test Results
Appearance | Silver grey Rods |
Composition | alloy typically:Bi 50%; Pb 25%; Cd 12.5%; Sn 12.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...