Sodium mercury amalgam Na ∼20%

Supplier: Thermo Scientific Chemicals
Danger

388460100 388460500
TS388460100EA 171 USD
TS388460100 TS388460500
Sodium mercury amalgam Na ∼20%
Sodium mercury amalgam (Mercury Sodium alloy (80:20 w%))
Formula: Hg:Na
Storage Temperature: Ambient
MDL Number: MFCD00210362
CAS Number: 11110-52-4
UN: 3401
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Silver to grey Irregular pieces
Sodium (Na) 20 % (approximately)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR