1-(4-Bromobutoxy)-4-nitrobenzene 85%
Supplier: Thermo Scientific Chemicals
|
Warning
|
385300050
385300250
TS385300050EA
47
USD
TS385300050
TS385300250
1-(4-Bromobutoxy)-4-nitrobenzene 85%
1-(4-Bromobutoxy)-4-nitrobenzene
Formula:
C₁₀H₁₂BrNO₃ Boiling Pt: 165…170 °C (.5 mmHg) Melting Pt: 37…40 °C |
CAS Number:
55502-03-9 |
Specification Test Results
Appearance | Beige to brown Adhering chunks |
Infrared spectrum | Conforms |
GC | ≥80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...