2,3,6-Trifluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

384180010 384180050
TS384180010EA 37 USD
TS384180010 TS384180050
2,3,6-Trifluorobenzaldehyde 97%
2,3,6-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 68…70 °C (10 mmHg)
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061195
CAS Number: 104451-70-9
UN: 2810

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4890 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR