Zinc chloride, anhydrous 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

380130050 380130250
TS380130050EA 73 USD
TS380130050 TS380130250
Zinc chloride, anhydrous 99.99% (trace metals basis)
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C (760 mmHg)
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR