3,5-Dibromo-2-pyridone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dibromo-2-hydroxypyridine, 3,5-Dibromo-2-pyridinol

379670050 379670010
TS379670050EA 83 USD
TS379670050 TS379670010
3,5-Dibromo-2-pyridone 98%
3,5-Dibromo-2-pyridone
Formula: C₅H₃Br₂NO
MW: 252.89 g/mol
Melting Pt: 204…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00023472
CAS Number: 13472-81-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 204 to 210 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR