2-Chloro-3-picoline 98%

Supplier: Thermo Scientific Chemicals
Warning

379580050 379580250
TS379580050EA 66 USD
TS379580050 TS379580250
2-Chloro-3-picoline 98%
2-Chloro-3-picoline
Formula: C₆H₆ClN
Boiling Pt: 192…193 °C (760 mmHg)
Density: 1.17 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00234177
CAS Number: 18368-76-8
EINECS: 242-242-1

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5340 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR