(4-Tolyloxy)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(p-Tolyloxy)acetic acid, 4-Methylphenoxyacetic acid, (4-Methylphenoxy)acetic acid

378270050 378270250
TS378270050EA 20 USD
TS378270050 TS378270250
(4-Tolyloxy)acetic acid 99%
(4-Tolyloxy)acetic acid
Formula: (C₆H₄(CH₃))OCH₂COOH
MW: 166.18 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00014365
CAS Number: 940-64-7
EINECS: 213-374-7

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 139 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR