3-Ethyl-3-oxetanemethanol 96%

Supplier: Thermo Scientific Chemicals
Warning

377700010 377700050
TS377700010EA 23 USD
TS377700010 TS377700050
3-Ethyl-3-oxetanemethanol 96%
Formula: C₆H₁₂O₂
Boiling Pt: 96 °C (4 mmHg)
Density: 1.019 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00190143
CAS Number: 3047-32-3
EINECS: 221-254-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20 °C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR