5-(4-Chlorophenyl)furfural 95+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
375500010
375500050
TS375500010EA
20
USD
TS375500010
TS375500050
5-(4-Chlorophenyl)furfural 95+%
5-(4-Chlorophenyl)furfural
Formula:
C₁₁H₇ClO₂ Melting Pt: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00195947 CAS Number: 34035-03-5 |
Specification Test Results
Appearance | Light brown Needles |
Infrared spectrum | Conforms |
Melting point | 123 to 127 °C |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...