α-Bromo-3-chloro-5-fluorotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene, 3-Chloro-5-fluorobenzyl bromide

374700010 374700050
TS374700010EA 48 USD
TS374700010 TS374700050
α-Bromo-3-chloro-5-fluorotoluene 97%
α-Bromo-3-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Melting Pt: 25…30 °C
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystals or crystalline mass or liquid
Infrared spectrum Conforms
Melting point 25 to 30 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR