2-Chlorobenzyl mercaptan 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chlorobenzenemethanethiol, o-Chlorotoluene-α-thiol

373130050 373130250
TS373130050EA 27 USD
TS373130050 TS373130250
2-Chlorobenzyl mercaptan 98%
2-Chlorobenzyl mercaptan
Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 120…121 °C (25 mmHg)
Density: 1.22 g/cm³
Flash Pt: 69 °C
MDL Number: MFCD00004868
CAS Number: 39718-00-8
EINECS: 254-602-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5890 to 1.5910 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR