1-(3-Fluorophenyl)-2-thiourea 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(3-Fluorophenyl)thiourea, 3-Fluorophenylthiourea

372510010 372510050
TS372510010EA 21 USD
TS372510010 TS372510050
1-(3-Fluorophenyl)-2-thiourea 98%
1-(3-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00041167
CAS Number: 458-05-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR