Hydroxylapatite for analysis

Supplier: Thermo Scientific Chemicals

371260010 371260250 371261000
TS371260010EA 167 USD
TS371260010 TS371260250 TS371261000
Hydroxylapatite for analysis
Hydroxylapatite
Formula: Ca₅HO₁₃P₃
Melting Pt: 1700 °C
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS Number: 1306-06-5
EINECS: 215-145-7

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 35 to 40 % (Ca)
Chloride (Cl) ≤1500 ppm
Fluoride (F) ≤50 ppm
Sulfate (SO4) ≤5000 ppm
Arsenic (As) ≤2 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤400 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm
Zinc (Zn) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition ≤8 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR