Sodium, dry stick
Supplier: Thermo Scientific Chemicals
|
Danger
|
370210010
370212500
TS370210010EA
460
USD
TS370210010
TS370212500
Sodium, dry stick
Sodium
Formula:
Na Boiling Pt: 883 °C Melting Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: 4.3,I Merck Index: 12,08710 |
Specification Test Results
Appearance | Light grey or silver white Sticks |
Calcium (Ca) | ≤400 ppm |
Potassium (K) | ≤300 ppm |
Sodium (Na) | ≥99.8 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...