4'-Fluoro-3'-nitroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

366010050 366010250
TS366010050EA 64 USD
TS366010050 TS366010250
4'-Fluoro-3'-nitroacetophenone 98%
4'-Fluoro-3'-nitroacetophenone
Formula: C₈H₆FNO₃
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00115369
CAS Number: 400-93-1

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 46 to 50 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR