3,5-Difluoro-2-methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid

365820010 365820050
TS365820010EA 47 USD
TS365820010 TS365820050
3,5-Difluoro-2-methoxyphenylboronic acid 97%
3,5-Difluoro-2-methoxyphenylboronic acid
Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD03095353
CAS Number: 737000-76-9

Order Now

Specification Test Results

Appearance Beige Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR