(2,3-Dichlorophenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

365550050 365550250
TS365550050EA 45 USD
TS365550050 TS365550250
(2,3-Dichlorophenyl)acetic acid 97%
(2,3-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD01861393
CAS Number: 10236-60-9

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 130 to 135 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR